Learn More
Sigma Organic Chemistry 1H-Pyrrolo 2 3-cpyridin | 1G | 271-29-4 | MFCD01686899
Supplier: Sigma Organic Chemistry CDS0075141G
1H-Pyrrolo 2 3-cpyridin , 1G
About this Item:
CAS #: 271-29-4
MDL #: MFCD01686899
Molecular Weight: 118.14
UNSPSC Code: 12352200
Chemical Formula: C7H6N2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.