Learn More
ALADDIN SCIENTIFIC CORPORATION 1Ha1Ha2Ha2H-Perfluorodecyltrie, 25g
Supplier:  ALADDIN SCIENTIFIC CORPORATION P12238525G
Encompass_Preferred
1H1H2H2H-Perfluorodecyltriethoxysilane(PFDTES) is a fluoroalkylsilanes(FAS) with low surface energy and high wettability due to the presence of fluorinated carbons It is used as a superhydrophobic agent for synthesizing various functional coatings A chemical to bond poly (tetrafluoroethylene) films to silicon wafers
Catalog No. 50-000-82558