Learn More
Leica Biosystems 1ml NCL-L-a-SARC
Supplier:  Leica Biosystems ASARCLCE
Encompass
Antigen Background In normal skeletal muscle, dystrophin, the protein product of the gene which is defective in Duchenne and Becker muscular dystrophy, is attached to the muscle membrane via a complex of proteins (dystrophin-associated glycoproteins, DAGs). Dystrophin-deficient muscle shows a generalized reduction in DAG labeling. The expression of different members of the dystrophin glycoprotein complex is altered in several types of muscular dystrophy. For example, patients with LGMD2D have mutations in the gene for alpha-sarcoglycan, those with LGM2E have mutations in the beta-sarcoglycan gene, those with LGM2C have mutations in the gamma-sarcoglycan gene and those with LGM2F have mutations in the delta-sarcoglycan gene. As the sarcoglycans function together as a sub-complex, mutations in any one of the sarcoglycan genes usually results in variable expression for the whole group. Disclaimer Sarcoglycan Antibodies are recommended for the detection of specific antigens of interest in
Catalog No. 50-255-2378
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.