Learn More
Leica Biosystems 1ml NCL-L-CD141
Supplier:  Leica Biosystems CD141LU
Encompass
Antigen Background Thrombomodulin is a transmembrane glycoprotein of 75 kD which can accelerate the activation of protein C. Activated protein C functions as an anticoagulant by combining with protein S to inactivate factors Va and VIIIa of the blood coagulation pathway and by binding thrombin. Several factors regulate thrombomodulin expression. Downregulation of thrombomodulin may be induced by the cytokine interleukin-1, tumor necrosis factor and endotoxin. Agents which increase cyclic AMP such as forskolin may upregulate thrombomodulin activity in endothelial cells. Thrombomodulin has been identified within a number of normal tissues. These include the lining cells of arteries, veins, capillaries and the lymphatics as well as mesothelial cells, meningeal lining cells, synovial cells, syncytiotrophoblasts, megakaryocytes and platelets. Disclaimer CD141 (Thrombomodulin) is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct t
Catalog No. 50-255-2337
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.