Learn More
Leica Biosystems 1ml NCL-L-CD146
Supplier:  Leica Biosystems CD146LU
Encompass
Antigen Background CD146 protein is also known as the melanoma metastasis-associated surface molecule, MUC18, A32 antigen, S-Endo-1 and the melanoma cell adhesion molecule, MCAM or Mel-CAM. Originally, the CD146 molecule was defined as a marker of tumor progression and metastasis formation in human melanoma. More recently, it has been reported to be expressed on endothelial cells, smooth muscle and cerebellar cortex. Structurally, CD146 is an integral membrane glycoprotein of 113 kD with the characteristic V-V-C2-C2-C2 immunoglobulin-like domain structure. It shares considerable homology with chicken neural adhesion molecule, chicken gicerin, goldfish neurolin and is also closely related to the human blood group glycoprotein, lutheran. Although CD146 molecule functions as a cell adhesion molecule it interacts with an as yet uncharacterized ligand. CD146 can be induced on all T cells via PHA, recall antigen, superantigen and T cell receptor/CD3 stimulation. Furthermore reports suggest t
Catalog No. 50-255-2336
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.