Learn More
Leica Biosystems 1ml NCL-L-CD5-4C7
Supplier:  Leica Biosystems CD54C7LU
Encompass
Antigen BackgroundCD5 antigen is reported to be expressed on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed on T cells. CD5 antigen is also expressed by many T cell leukemias, lymphomas, activated T cells and on a subset of B cells located primarily in the mantle zones of normal lymph nodes. CD5 antigen expression is also reported in T cell acute lymphocytic leukemias (T-ALL), some B cell chronic lymphocytic leukemias (B-CLL) as well as B and T cell lymphomas.DisclaimerCD5 is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2307
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.