Learn More
Leica Biosystems 1ml NCL-L-IgM
Supplier:  Leica Biosystems IGMLCE
Encompass
Antigen Background IgM, together with IgD, is the major immunoglobulin expressed on the surface of B cells and normally constitutes about 10 per cent of serum immunoglobulin. IgM antibody is prominent in early immune responses to most antigens and predominates in certain antibody responses such as natural blood group antibodies. Disclaimer Immunoglobulin M is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2230
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.