Learn More
Leica Biosystems 1ml NCL-L-NF200-N52
Supplier:  Leica Biosystems NF200N52LCE
Encompass
Antigen Background Neurofilaments constitute the main structural elements of neuronal axons and dendrites. Neurofilaments are composed of three major subunits referred to as the neurofilament triplet, with molecular weights of 68 kD, 160kD and 200 kD. Within tumors, only neoplastic cells of neural origin or those exhibiting neuronal differentiation, have been reported to express neurofilaments. Disclaimer Neurofilament 200kD is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2197
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.