Learn More
Leica Biosystems 1ml NCL-L-TOPO I
Supplier:  Leica Biosystems TOPOILU
Encompass
Antigen Background Topoisomerases are nuclear enzymes involved in a variety of cellular activities such as chromosomal condensation, DNA replication, transcription, recombination and segregation at mitosis. Human Topoisomerase I is a 100 kD protein capable of relaxing positively and negatively supercoiled DNA by performing a transient single-stranded nick which is then re-ligated at the end of the reaction. It has been shown that the enzyme is located in regions of the genome that are undergoing active RNA synthesis where it probably reduces superhelical stresses in the DNA enabling RNA polymerase to function properly. In normal eukaryotic cells, DNA topoisomerase I does not show relevant fluctuations across the cell cycle, unlike DNA topoisomerase II alpha. Both DNA topoisomerases I and II have been found to be targets of autoantibodies in the sera of individuals with certain autoimmune diseases, for example, systemic lupus erythematosus and also of some anti-tumor drugs and antibioti
Catalog No. 50-255-1960
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.