Learn More
Leica Biosystems 1ml NCL-L-TPO
Supplier:  Leica Biosystems TPOLCE
Encompass
Antigen Background Thyroid peroxidase gene expression is under the regulation of thyroid stimulating hormone. In normal thyroid, expression of thyroid peroxidase (TPO) described immunohistochemically is reported to produce a diffuse, fine, granular cytoplasmic stain in all follicular cells. Some studies have shown qualitative, as well as quantitative differences in thyroid peroxidase expression in thyroid cancer compared to normal tissue. Product Specific Information TPO stains optimally when used in TBS-based wash buffer and diluent systems. Disclaimer Thyroid Peroxidase is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-1958
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.