Learn More
Leica Biosystems 1ml NCL-L-TSH
Supplier:  Leica Biosystems TSHLU
Encompass
Antigen Background Thyroid stimulating hormone (TSH) is a pituitary hormone of 28 kD which stimulates thyroid growth and production of thyroid hormones. TSH is reported to be expressed in thyrotrophic cells of the pituitary and pituitary adenomas. Disclaimer Thyroid Stimulating Hormone is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-1956
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.