Learn More
Leica Biosystems 1ml NCL-L-WAF-1
Supplier:  Leica Biosystems WAF1L
Encompass
Antigen Background The gene encoding WAF1, also termed p21, is transcriptionally regulated by the suppressor protein, p53. Overexpression of WAF1 is growth suppressive, possibly by inhibiting the activity of cyclin/CDK complexes. One consequence of WAF1 binding to cyclin/CDK is the inhibition of Rb protein phosphorylation. Induction of WAF1 expression requires wild type p53 activity in cells undergoing p53 dependent G1 arrest or apoptosis. Mutation of the p53 gene is a common event in human cancer and results in the failure to produce WAF1. The effect of this may lead to uncontrolled cell proliferation. Disclaimer p21 (WAF1 Protein) is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-1942
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.