Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2R)-(-)-N,N'-Dimethylcyclohexane-1,2-diamine 94.0+%, TCI America™
SDP

Catalog No. D2459100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2459100MG 100 mg
D24591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2459100MG Supplier TCI America Supplier No. D2459100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 68737-65-5
Molecular Formula C8H20N2
Molecular Weight (g/mol) 144.26
MDL Number MFCD00671527,MFCD03001702
InChI Key JRHPOFJADXHYBR-HTQZYQBOSA-P
Synonym 1r,2r-n,n'-dimethyl-1,2-cyclohexanediamine, 1r,2r-n1,n2-dimethylcyclohexane-1,2-diamine, 1r,2r---n,n'-dimethylcyclohexane-1,2-diamine, trans-n1,n2-dimethylcyclohexane-1,2-diamine, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine, trans-n,n'-dimethylcyclohexane-1,2-diamine, trans-n,n'-dimethyl-1,2-cyclohexanediamine, trans-1,2-bis methylamino cyclohexane, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexane diamine, 1r,2r---1,2-bis methylamino cyclohexane
PubChem CID 2733821
IUPAC Name (1R,2R)-N1,N2-dimethylcyclohexane-1,2-bis(aminium)
SMILES C[NH2+][C@@H]1CCCC[C@H]1[NH2+]C

Specifications

Melting Point 45°C
Quantity 100 mg
UN Number 3259
Formula Weight 142.25
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2R)-(-)-N,N′-Dimethylcyclohexane-1,2-diamine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.