Learn More
Medchemexpress LLC (1R,2S)-Xeruborbactam disodium | 2170836-14-1 | 99.3% | 283.96 g/mol | C10H8BFNa2O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY136072A5MG
Encompass_Preferred
(1R,2S)-Xeruborbactam disodium is a boronic acid-based beta-lactamase inhibitor supplied as a high-purity powdered disodium salt for research use. It is intended for biochemical and microbiological studies of beta-lactamase inhibition and for investigations of antibiotic potentiation.
  • High purity suitable for analytical and bioassay applications.
  • Powder form for easy aliquoting and storage.
  • Useful in assays targeting serine and metallo beta-lactamases.
  • Provided in small-quantity vials for early-stage research.
  • Stable when stored under recommended cold conditions.
Catalog No. 50-002-22199