Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R)-Camphor Oxime 98.0+%, TCI America™
SDP

Catalog No. C00131G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C00131G 1 g
C00135G 5 g
C001325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C00131G Supplier TCI America Supplier No. C00131G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2792-42-9
Molecular Formula C10H17NO
Molecular Weight (g/mol) 167.25
MDL Number MFCD00046298,MFCD00210161
InChI Key OVFDEGGJFJECAT-DHZHZOJONA-N
PubChem CID 124925274
IUPAC Name N-[(2E)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-ylidene]hydroxylamine
SMILES CC1(C)C2CCC1(C)\C(C2)=N\O

Specifications

Melting Point 119°C
Color White
Quantity 1 g
Formula Weight 167.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (1R)-Camphor Oxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.