Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R)-(-)-Camphorquinone 98.0+%, TCI America™
SDP

Catalog No. C14821G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14821G 1 g
C14825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14821G Supplier TCI America Supplier No. C14821G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10334-26-6
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00064160,MFCD00082863,MFCD00064160,MFCD00082863
InChI Key VNQXSTWCDUXYEZ-LDWIPMOCSA-N
Synonym 1r,4s-1,7,7-trimethylnorbornane-2,3-dione, 1r,4r-4,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione, 1s,4r-4,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione
PubChem CID 16211789
IUPAC Name (1R,4S)-1,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
SMILES CC1(C)[C@@H]2CC[C@@]1(C)C(=O)C2=O

Specifications

Melting Point 200°C
Color Yellow
Quantity 1 g
Formula Weight 166.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (1R)-(-)-Camphorquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.