Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,2S)-(-)-1,2-Cyclohexanediamine D-Tartrate 98.0+%, TCI America™
SDP

Catalog No. C145025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C145025G 25 g
C14505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C145025G Supplier TCI America Supplier No. C145025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67333-70-4
Molecular Formula C10H20N2O6
Molecular Weight (g/mol) 264.278
MDL Number MFCD00191980
InChI Key GDOTUTAQOJUZOF-SCDVTJNCSA-N
Synonym 1s,2s---1,2-diaminocyclohexane d-tartrate, 1s,2s-+-cyclohexane-1,2-diamine d-tartrate salt, 1s,2s---1,2-cyclohexanediamine d-tartrate, 1s,2s---1,2-diaminocyclohexane l-tartrate, 1s,2s-cyclohexane-1,2-diamine 2s,3s-2,3-dihydroxysuccinate, 1s,2s---cyclohexane-1,2-diamine d-tartrate, 1s,2s-1,2-diaminocyclohexane d-tartrate, 1s,2s---1,2-diaminocyclohexane-d-tartrate, 1s,2s-cyclohexane-1,2-diamine; 2s,3s-2,3-dihydroxybutanedioic acid, 1,2-cyclohexanediamine, 1s,2s-, 2s,3s-2,3-dihydroxybutanedioate 1:1
PubChem CID 16212699
IUPAC Name (1S,2S)-cyclohexane-1,2-diamine;(2S,3S)-2,3-dihydroxybutanedioic acid
SMILES C1CCC(C(C1)N)N.C(C(C(=O)O)O)(C(=O)O)O

Specifications

Melting Point 284°C
Color White
Quantity 25 g
Formula Weight 264.28
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (1S,2S)-(-)-1,2-Cyclohexanediamine D-Tartrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.