Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S)-3-(1,10-Phenanthrolin-2-yl)-2'-phenyl-[1,1'-binaphthalen]-2-ol 97.0+%, TCI America™
SDP

Catalog No. P259920MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P259920MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P259920MG Supplier TCI America Supplier No. P259920MG

Chemical Identifiers

CAS 1621903-81-8
Molecular Formula C38H24N2O
Synonym (S)-BinThro

Specifications

Color White-Yellow
Quantity 20 mg
Formula Weight 524.62
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (1S)-3-(1,10-Phenanthrolin-2-yl)-2′-phenyl-[1,1′-binaphthalen]-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.