Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S)-(-)-Camphanic chloride, 95%

Catalog No. AAL1414806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1414806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1414806 Supplier Thermo Scientific Chemicals Supplier No. L1414806

missing translation for 'validMainframeMsg'

CAS: 39637-74-6 | C10H13ClO3 | 216.661 g/mol

(1S)-(-)-Camphanic chloride is used as a resolving agent for alcohols as the diastereomeric esters by crystallization or chromatography and as a chiral derivatization reagent for determination of enantiomeric excess of alcohols and amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1S)-(-)-Camphanic chloride is used as a resolving agent for alcohols as the diastereomeric esters by crystallization or chromatography and as a chiral derivatization reagent for determination of enantiomeric excess of alcohols and amines.

Solubility
Soluble in dichloromethane, ether, ethyl acetate and methanol. Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C. Store under inert gas. It is sensitive to moisture. Protect from humidity and water. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39637-74-6
Molecular Formula C10H13ClO3
Molecular Weight (g/mol) 216.661
MDL Number MFCD00135626
InChI Key PAXWODJTHKJQDZ-QVDQXJPCSA-N
Synonym 1s-4,7,7-trimethyl-3-oxo-2-oxabicyclo 2.2.1 heptane-1-carbonyl chloride, --1s,4r-camphanoyl chloride, 1s---camphanic acid chloride
PubChem CID 12364938
IUPAC Name (4S)-1,7,7-trimethyl-2-oxo-3-oxabicyclo[2.2.1]heptane-4-carbonyl chloride
SMILES CC1(C2(CCC1(OC2=O)C(=O)Cl)C)C

Specifications

Melting Point 64°C to 68°C
Quantity 5 g
UN Number UN3261
Beilstein 3590860
Sensitivity Moisture sensitive
Solubility Information Soluble in dichloromethane,ether,ethyl acetate and methanol. Reacts with water.
Optical Rotation −18° (c=2 in Carbon Tetrachloride)
Formula Weight 216.67
Percent Purity 95%
Chemical Name or Material (1S)-(-)-Camphanic chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 254-552-4
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.