Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S)-(+)-Menthyl acetate, ≥99.0% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

Catalog No. 111016032 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6032 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6032 Supplier MilliporeSigma Supelco Supplier No. 459875MLF

Specifications

CAS 99-99-0
Density 0.925 g/mL (at 25°C)
Flash Point 77°C
Physical Form Neat
Linear Formula C12H22O2
MDL Number MFCD00062977
Boiling Point 228°C to 229°C
Type Analytical Standard
Quantity 5 mL
Formula Weight 198.30
Percent Purity ≥99.0% (sum of enantiomers, GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.