Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ 1SCAP2 Recombinant Protein (P02)

Catalog No. 89011439 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-011-439 10 μg
89-011-440 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-011-439 Supplier Abnova™ Supplier No. H00008935P0210

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Molecular Weight (g/mol) 65.23
Name Human SCAP2 Full-length ORF Recombinant Protein with GST-tag at N-terminal
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE stained with Coomassie Blue
Quantity 10 μg
Source Wheat Germ (in vitro)
Storage Requirements -80°C
Cross Reactivity Human
Recombinant Recombinant
Form Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.