Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,1,3-Benzoselenadiazole 98.0+%, TCI America™
SDP

Catalog No. B43131G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43131G 1 g
B4313200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43131G Supplier TCI America Supplier No. B43131G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 273-15-4
Molecular Formula C6H4N2Se
Molecular Weight (g/mol) 183.083
MDL Number MFCD00022886
InChI Key AYTPIVIDHMVGSX-UHFFFAOYSA-N
PubChem CID 67503
IUPAC Name 2,1,3-benzoselenadiazole
SMILES C1=CC2=N[Se]N=C2C=C1

Specifications

Melting Point 75°C
Boiling Point 246°C
Quantity 1 g
UN Number 3283
Formula Weight 183.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,1,3-Benzoselenadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.