Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(1,3-Dioxan-2-yl)ethyltriphenylphosphonium Bromide 98.0+%, TCI America™
SDP

Catalog No. D165525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D165525G 25 g
D16555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D165525G Supplier TCI America Supplier No. D165525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 69891-92-5
Molecular Formula C24H26BrO2P
Molecular Weight (g/mol) 457.35
MDL Number MFCD00012000
InChI Key XETDBHNHTOJWPZ-UHFFFAOYSA-M
Synonym 2-1,3-dioxan-2-yl ethyltriphenylphosphonium bromide, 2-1,3-dioxan-2-yl ethyl triphenylphosphonium bromide, 2-1,3-dioxan-2-yl ethyl triphenylphosphanium bromide, acmc-209obj, xqaupnaycikwct-uhfffaoysa-m, 1,3-dioxan-2-ylethyltriphenylphosphinebromide, 2-1,3-dioxan-2-yl ethyltriphenylphosphonium bromi, 2-1,3-dioxan-2-yl ethyltriphenylphosphoniumbromide, 2-1,3-dioxane-2-yl ethyltriphenylphosphonium bromide
PubChem CID 2724168
IUPAC Name [2-(1,3-dioxan-2-yl)ethyl]triphenylphosphanium bromide
SMILES [Br-].C(C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C1OCCCO1

Specifications

Melting Point 189°C
Color White-Yellow
Quantity 25 g
Formula Weight 457.35
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(1,3-Dioxan-2-yl)ethyltriphenylphosphonium Bromide
EINECSNumber (5)-6416
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.