Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(1-Azepanyl)-6-bromobenzonitrile, 96%, Thermo Scientific™

Catalog No. AAH6121603
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6121603 1 g
AAH61216MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6121603 Supplier Thermo Scientific Chemicals Supplier No. H6121603

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1260805-87-5
Molecular Formula C13H15BrN2
Molecular Weight (g/mol) 279.181
InChI Key PFOLTHPOEJJYAZ-UHFFFAOYSA-N
Synonym 2-azepan-1-yl-6-bromobenzonitrile
PubChem CID 75360876
IUPAC Name 2-(azepan-1-yl)-6-bromobenzonitrile
SMILES C1CCCN(CC1)C2=C(C(=CC=C2)Br)C#N

Specifications

Quantity 1 g
UN Number UN3439
Formula Weight 279.18
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-(1-Azepanyl)-6-bromobenzonitrile
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.