Learn More
Sigma Organic Chemistry 2- 1-Naphthyl propan-2- | 1G | MFCD11841274
Supplier: Sigma Organic Chemistry CBR008161G

2- 1-Naphthyl propan-2- , 1G
About this Item:
MDL #: MFCD11841274
Molecular Weight: 221.73
UNSPSC Code: 12352200
Chemical Formula: C13H16ClN
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.