Learn More
Medchemexpress LLC 2(1H)-pyrimidinone, 6-amino-5-bromo- | 2240-25-7 | MFCD00056025 | 99.8% | 190.00 g/mol | C4H4BrN3O | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0152055G
Encompass_Preferred
5-Bromocytosine is a brominated cytosine derivative supplied for laboratory research. It can be incorporated into cellular DNA and promotes DNA chain breakage and cross-linking under ultraviolet irradiation, making it useful for studies of DNA damage and repair mechanisms.
  • Brominated cytosine derivative used to study DNA incorporation and UV-induced DNA damage.
  • High purity: 99.77% (HPLC) as reported on the certificate of analysis.
  • Chemical formula C4H4BrN3O and molecular weight 190.00 g/mol.
  • Available in laboratory-scale pack sizes, including a 5 g option.
  • Provided with certificate of analysis containing batch-specific data.
  • For research use only; consult the safety data sheet for handling and storage.
Catalog No. 50-003-03377