Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[2-(2-Chloroethoxy)ethoxy]ethanol 96.0+%, TCI America™
SDP

Catalog No. C141625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C141625G 25 g
C1416100G 100 g
C1416500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C141625G Supplier TCI America Supplier No. C141625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5197-62-6
Molecular Formula C6H13ClO3
Molecular Weight (g/mol) 168.617
MDL Number MFCD00002874
InChI Key KECMLGZOQMJIBM-UHFFFAOYSA-N
PubChem CID 78871
IUPAC Name 2-[2-(2-chloroethoxy)ethoxy]ethanol
SMILES C(COCCOCCCl)O

Specifications

Color Yellow
Boiling Point 232°C
Quantity 25 g
Synonym 2-2-2-chloroethoxy ethoxy ethanol, ethanol, 2-2-2-chloroethoxy ethoxy, triethylene glycol monochloride, triethylene glycol monochlorohydrin, ethanol,2-2-2-chloroethoxy ethoxy, 2-2-2-chloro-ethoxy ethoxy ethanol, 2-2-2-chloroethoxy ethoxy-ethanol
Formula Weight 168.62
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-[2-(2-Chloroethoxy)ethoxy]ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.