Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trichloroethanol, 99%

Catalog No. AAL0816314
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0816314 25 g
AAL0816306 5 g
AAL0816322 100 g
AAL0816336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAL0816314 Supplier Thermo Scientific Chemicals Supplier No. L0816314
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 115-20-8 | C2H3Cl3O | 149.395 g/mol

2,2,2-Trichloroethanol is used as a sedative or a hypnotic. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trichloroethanol is used as a sedative or a hypnotic. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Store away from water and oxidizing agents. Protect from humidity and water. Incompatible with oxidizing agents, Strong acids, Strong bases, Strong reducing agents, Acid chlorides, Acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-20-8
Molecular Formula C2H3Cl3O
Molecular Weight (g/mol) 149.395
MDL Number MFCD00004677
InChI Key KPWDGTGXUYRARH-UHFFFAOYSA-N
Synonym trichloroethanol, trichlorethanol, trichloroethyl alcohol, ethanol, 2,2,2-trichloro, 2,2,2-trichloro-1-ethanol, hydroxymethyl trichloromethane, 2,2,2-trichloroethyl alcohol, 2,2,2-trichloroethan-1-ol, unii-aw835aj62n, beta-trichloroethanol
PubChem CID 8259
ChEBI CHEBI:28094
IUPAC Name 2,2,2-trichloroethanol
SMILES C(C(Cl)(Cl)Cl)O

Specifications

Melting Point 17°C to 18°C
Density 1.56
pH 5 to 6
Boiling Point 151°C to 153°C
Flash Point >110°C (230°F)
Refractive Index 1.4895
Quantity 25 g
Beilstein 1697495
Sensitivity Hygroscopic
Merck Index 14,9638
Solubility Information Slightly soluble in water.
Formula Weight 149.4
Percent Purity 99%
Chemical Name or Material 2,2,2-Trichloroethanol
Show More Show Less
Hazard Category H227-H302-H315-H318-H336
Hazard Statement GHS H Statement
H318-H302-H336-H315
Causes serious eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.
Causes skin irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 204-071-0
RTECSNumber KM3850000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.