Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trichloroethanol 98.0+%, TCI America™
SDP

Catalog No. T0622100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0622100G 100 g
T062225G 25 g
T0622500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0622100G Supplier TCI America Supplier No. T0622100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 115-20-8
Molecular Formula C2H3Cl3O
Molecular Weight (g/mol) 149.395
MDL Number MFCD00004677
InChI Key KPWDGTGXUYRARH-UHFFFAOYSA-N
Synonym trichloroethanol, trichlorethanol, trichloroethyl alcohol, ethanol, 2,2,2-trichloro, 2,2,2-trichloro-1-ethanol, hydroxymethyl trichloromethane, 2,2,2-trichloroethyl alcohol, 2,2,2-trichloroethan-1-ol, unii-aw835aj62n, beta-trichloroethanol
PubChem CID 8259
ChEBI CHEBI:28094
IUPAC Name 2,2,2-trichloroethanol
SMILES C(C(Cl)(Cl)Cl)O

Specifications

Melting Point 18°C
Color Yellow
Boiling Point 151°C
Quantity 100 g
UN Number 1760
Formula Weight 149.40
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,2-Trichloroethanol
EINECSNumber (2)-2002
RTECSNumber KM3850000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.