Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trichloroethyl Chloroformate 98.0+%, TCI America™
SDP

Catalog No. C0795250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0795250G 250 g
C079525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0795250G Supplier TCI America Supplier No. C0795250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17341-93-4
Molecular Formula C3H2Cl4O2
Molecular Weight (g/mol) 211.847
MDL Number MFCD00000810
InChI Key LJCZNYWLQZZIOS-UHFFFAOYSA-N
Synonym 2,2,2-trichloroethyl chloroformate, trichloroethyl chloroformate, 2,2,2-trichloroethylchloroformate, carbonochloridic acid, 2,2,2-trichloroethyl ester, 2,2,2-trichloroethoxycarbonyl chloride, chloroformic acid 2,2,2-trichloroethyl ester, formic acid, chloro-, 2,2,2-trichloroethyl ester, chloro 2,2,2-trichloroethoxy methanone, beta, beta, beta-trichloroethoxycarbonyl chloride, troc
PubChem CID 87063
IUPAC Name 2,2,2-trichloroethyl carbonochloridate
SMILES C(C(Cl)(Cl)Cl)OC(=O)Cl

Specifications

Color Colorless
Boiling Point 172°C
Quantity 250 g
UN Number 3277
Formula Weight 211.85
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,2-Trichloroethyl Chloroformate
EINECSNumber (2)-3319
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.