Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoroethanol, For NMR spectroscopy, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018135 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8135 100 mL
11-101-8997 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8135 Supplier MilliporeSigma Supelco Supplier No. 91683100ML
Add to Cart
Add to Cart
Featured Industries: Environmental
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 896°F
CAS 75-89-8
Vapor Pressure 70 mmHg (25°C)
Vapor Density 3.5 (vs. air)
Density 1.373 g/mL (at 25°C)
Flash Point 29°C
Physical Form Neat
Linear Formula CF3 CH2 OH
MDL Number MFCD00004672
Melting Point -44°C
Boiling Point 77°C to 80°C
Synonym Trifluoroethyl alcohol
Type Analytical Standard
Quantity 100 mL
Formula Weight 100.04
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.