Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoroethyl Butyrate 98.0+%, TCI America™
SDP

Catalog No. B214425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B214425G 25 g
B21445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B214425G Supplier TCI America Supplier No. B214425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 371-27-7
Molecular Formula C6H9F3O2
Molecular Weight (g/mol) 170.131
MDL Number MFCD00077609
InChI Key DEXWRCYOMLUJRF-UHFFFAOYSA-N
PubChem CID 193290
IUPAC Name 2,2,2-trifluoroethyl butanoate
SMILES CCCC(=O)OCC(F)(F)F

Specifications

Color Colorless
Boiling Point 113°C
Quantity 25 g
UN Number 3272
Synonym 2,2,2-trifluoroethyl butyrate, 2,2,2-trifluoroethylbutyrate, butanoic acid, 2,2,2-trifluoroethyl ester, 2-tfb, butyric acid, tfe, trifluoroethyl butyrate, trifluoroethyl butanoate, acmc-209ir0, butanoic acid,2,2,2-trifluoroethyl ester
Formula Weight 170.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,2-Trifluoroethyl Butyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.