Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoroethyl Methanesulfonate 98.0+%, TCI America™
SDP

Catalog No. M119325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M119325G 25 g
M11935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M119325G Supplier TCI America Supplier No. M119325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25236-64-0
Molecular Formula C3H5F3O3S
Molecular Weight (g/mol) 178.13
MDL Number MFCD00025058
InChI Key ICECLJDLAVVEOW-UHFFFAOYSA-N
Synonym 2,2,2-trifluoromethyl mesylate, methanesulfonic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl methanesulphonate, 2,2,2-trifluoroethyl mesylate, 2,2,2-trifluoroethyl methylsulfonate, acmc-209gib, cf3ch2oso2ch3, icecljdlavveow-uhfffaoysa, 2,2,2-trifluoroethyl methansulfonate, 2,2,2 trifluoroethyl methanesulfonate
PubChem CID 556565
IUPAC Name 2,2,2-trifluoroethyl methanesulfonate
SMILES CS(=O)(=O)OCC(F)(F)F

Specifications

Color Colorless
Boiling Point 99°C
Quantity 25 g
UN Number 3265
Formula Weight 178.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,2-Trifluoroethyl Methanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.