Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoroethyl trichloromethanesulfonate, 94%

Catalog No. AAL0863503
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0863503 1 g
AAL0863506 5 g
AAL0863514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0863503 Supplier Thermo Scientific Chemicals Supplier No. L0863503
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 23199-56-6 | C3H2Cl3F3O3S | 281.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23199-56-6
Molecular Formula C3H2Cl3F3O3S
Molecular Weight (g/mol) 281.45
MDL Number MFCD00042400
InChI Key GOIWZZQXWJVDOG-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethyl trichloromethanesulphonate, 2,2,2-trifluoroethyl trichloromethane-sulfonate, trichloromethanesulfonic acid 2,2,2-trifluoroethyl ester, methanesulfonic acid, trichloro-, 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl trichloromethyl sulfonate, acmc-20aoku, trifluoroethyl trichloromethylsulfonate, trifluoroethyl trichloromethanesulphonate, 2,2,2-trifluoroethyltrichloromethanesulfonate, 1,1,1-trifluoroethyl trichloromethanesulfonate
PubChem CID 90027
IUPAC Name 2,2,2-trifluoroethyl trichloromethanesulfonate
SMILES FC(F)(F)COS(=O)(=O)C(Cl)(Cl)Cl

Specifications

Melting Point -70°C
Density 1.714
Boiling Point 84°C to 86°C (20 mmHg)
Odor Acrid
Refractive Index 1.422
Quantity 1 g
UN Number UN3265
Beilstein 1108366
Sensitivity Moisture sensitive
Formula Weight 281.47
Percent Purity 94%
Chemical Name or Material 2,2,2-Trifluoroethyl trichloromethanesulfonate
Show More Show Less
missing translation for 'healthHazard1' H314-H335
missing translation for 'healthHazard2' GHS H Statement

H314-H318

Causes severe skin burns and eye damage.

Causes serious eye damage.
missing translation for 'healthHazard3' P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 245-484-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.