Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trimethylacetophenone, 97%

Catalog No. AAL0872906
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0872906 5 g
AAL0872903 1 g
AAL0872914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0872906 Supplier Thermo Scientific Chemicals Supplier No. L0872906
Add to Cart

CAS: 938-16-9 | C11H14O | 162.232 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Not miscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 938-16-9
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.232
MDL Number MFCD00008844
InChI Key OECPUBRNDKXFDX-UHFFFAOYSA-N
Synonym 2,2-dimethylpropiophenone, pivalophenone, 2,2,2-trimethylacetophenone, phenyl tert-butyl ketone, tert-butyl phenyl ketone, 1-propanone, 2,2-dimethyl-1-phenyl, unii-96ba178unx, t-bubz, t-butyl phenyl ketone, dsstox_cid_28184
PubChem CID 70308
IUPAC Name 2,2-dimethyl-1-phenylpropan-1-one
SMILES CC(C)(C)C(=O)C1=CC=CC=C1

Specifications

Density 0.97
Boiling Point 219°C to 222°C
Flash Point 87°C (188°F)
Refractive Index 1.5084
Quantity 5 g
Beilstein 1906460
Solubility Information Not miscible with water.
Formula Weight 162.23
Percent Purity 98%
Chemical Name or Material 2,2,2-Trimethylacetophenone
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 213-338-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.