Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3,3,3-Pentafluoropropyl p-Toluenesulfonate 98.0+%, TCI America™
SDP

Catalog No. P18345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18345G 5 g
P183425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18345G Supplier TCI America Supplier No. P18345G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 565-42-4
Molecular Formula C10H9F5O3S
Molecular Weight (g/mol) 304.23
MDL Number MFCD02093346
InChI Key JBHQQXONFHOEQU-UHFFFAOYSA-N
Synonym p-Toluenesulfonic Acid 2,2,3,3,3-Pentafluoropropyl Ester, 2,2,3,3,3-Pentafluoropropyl Tosylate
PubChem CID 2776017
IUPAC Name 2,2,3,3,3-pentafluoropropyl 4-methylbenzene-1-sulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC(F)(F)C(F)(F)F

Specifications

Melting Point 54°C
Color White-Yellow
Boiling Point 75°C
Quantity 5 g
Formula Weight 304.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2,3,3,3-Pentafluoropropyl p-Toluenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.