Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3,3,4,4,4-Heptafluoro-1-butanol 98.0+%, TCI America™
SDP

Catalog No. H054825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H054825G 25 g
H05485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H054825G Supplier TCI America Supplier No. H054825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 375-01-9
Molecular Formula C4H3F7O
Molecular Weight (g/mol) 200.06
MDL Number MFCD00004674
InChI Key WXJFKAZDSQLPBX-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4,4-heptafluoro-1-butanol, 1-butanol, 2,2,3,3,4,4,4-heptafluoro, 1h,1h-heptafluorobutanol-1, 1h,1h-heptafluoro-1-butanol, 1,1-dihydroperfluorobutyl alcohol, 2,2,3,3,4,4,4-heptafluorobutanol, 1,1-dihydroperfluorobutanol, 1,1-h,h-heptafluorobutanol, perfluoro-1,1-dihydrobutanol, butanol, 2,2,3,3,4,4,4-heptafluoro
PubChem CID 9776
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutan-1-ol
SMILES OCC(F)(F)C(F)(F)C(F)(F)F

Specifications

Color Yellow
Boiling Point 96°C
Quantity 25 g
UN Number 1987
Formula Weight 200.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3,3,4,4,4-Heptafluoro-1-butanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.