Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl Methacrylate (stabilized with TBC) 97.0+%, TCI America™
SDP

Catalog No. D499025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D499025G 25 g
D49905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D499025G Supplier TCI America Supplier No. D499025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2261-99-6
Molecular Formula C11H8F12O2
Molecular Weight (g/mol) 400.164
MDL Number MFCD00080658
InChI Key YJKHMSPWWGBKTN-UHFFFAOYSA-N
Synonym Methacrylic Acid 2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl Ester
PubChem CID 102236
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoroheptyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(C(C(C(C(C(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Yellow
Boiling Point 112°C
Quantity 25 g
Formula Weight 400.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl Methacrylate (stabilized with TBC)
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.