Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3,3,4,4,5,5-Octafluoro-1-pentanol 98.0+%, TCI America™
SDP

Catalog No. O0114100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0114100G 100 g
O011425G 25 g
O0114500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O0114100G Supplier TCI America Supplier No. O0114100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 355-80-6
Molecular Formula C5H4F8O
Molecular Weight (g/mol) 232.07
MDL Number MFCD00039631
InChI Key JUGSKHLZINSXPQ-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4,5,5-octafluoro-1-pentanol, 1h,1h,5h-octafluoro-1-pentanol, 1-pentanol, 2,2,3,3,4,4,5,5-octafluoro, 1,1,5-trihydrooctafluoropentyl alcohol, 2,2,3,3,4,4,5,5-octafluoropentanol, 1,1,5-trihydrooctafluoropentan-1-ol, unii-d05k8ejx15, 1h,1h,5h-octafluoropentanol, 2,2,3,3,4,4,5,5-octafluoropentyl alcohol, alpha,alpha,omega-trihydroperfluoropentanol
PubChem CID 9641
IUPAC Name 2,2,3,3,4,4,5,5-octafluoropentan-1-ol
SMILES OCC(F)(F)C(F)(F)C(F)(F)C(F)F

Specifications

Color Colorless
Boiling Point 140°C
Quantity 100 g
Formula Weight 232.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3,3,4,4,5,5-Octafluoro-1-pentanol
RTECSNumber SA7650000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.