Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3,3-Tetramethoxybutane, 95%

Catalog No. AC450930010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450930010 1 g
AC450930050 5 g
AC450930250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC450930010 Supplier Thermo Scientific Chemicals Supplier No. 450930010

CAS: 176798-33-7 | C8H18O4 | 178.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 176798-33-7
Molecular Formula C8H18O4
Molecular Weight (g/mol) 178.23
MDL Number MFCD25458126
InChI Key COEBCOIFCLAOFZ-UHFFFAOYSA-N
Synonym butane, 2,2,3,3-tetramethoxy
PubChem CID 10856041
IUPAC Name 2,2,3,3-tetramethoxybutane
SMILES COC(C)(OC)C(C)(OC)OC

Specifications

Boiling Point 172.0°C to 174.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 178.23
Percent Purity 95%
Chemical Name or Material 2,2,3,3-Tetramethoxybutane
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Flammable liquid and vapour.
Precautionary Statement GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.