Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3,4,4,4-Hexafluorobutyl Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™
SDP

Catalog No. H1555100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1555100G 100 g
H15555G 5 g
H155525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H1555100G Supplier TCI America Supplier No. H1555100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 36405-47-7
Molecular Formula C8H8F6O2
Molecular Weight (g/mol) 250.14
MDL Number MFCD00042311
InChI Key DFVPUWGVOPDJTC-UHFFFAOYSA-N
Synonym 2,2,3,4,4,4-hexafluorobutyl methacrylate, 1h,1h,3h-hexafluorobutyl methacrylate, hexafluorobutyl methacrylate, methacrylic acid 2,2,3,4,4,4-hexafluorobutyl ester, 2-propenoic acid, 2-methyl-, 2,2,3,4,4,4-hexafluorobutyl ester, hexafluorobutyl methacrylate polymer, hfbma, acmc-1adv6, ksc489s1j, 2-propenoic acid, 2-methyl-, 2,2,3,4,4,4-hexafluorobutyl ester, homopolymer
PubChem CID 549772
IUPAC Name 2,2,3,4,4,4-hexafluorobutyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(C(C(F)(F)F)F)(F)F

Specifications

Color Colorless
Boiling Point 158°C
Quantity 100 g
UN Number 3272
Formula Weight 250.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3,4,4,4-Hexafluorobutyl Methacrylate (stabilized with MEHQ)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.