Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2,3-dichlorophenyl)-1,3-thiazole-4-carboxylic acid, Thermo Scientific™

Catalog No. SPB02181CB
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SPB02181CB 250 mg
SPB02181DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SPB02181CB Supplier Thermo Scientific Chemicals Supplier No. SPB02181CB
Add to Cart

Chemical Identifiers

CAS 257876-07-6
Molecular Formula C10H5Cl2NO2S
Molecular Weight (g/mol) 274.115
InChI Key ANTJCNIMRNBFMJ-UHFFFAOYSA-N
Synonym 2-2,3-dichlorophenyl-1,3-thiazole-4-carboxylic acid, 2-2,3-dichloro-phenyl-thiazole-4-carboxylic acid, 2-2,3-dichlorophenyl thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-2,3-dichlorophenyl
PubChem CID 2743987
IUPAC Name 2-(2,3-dichlorophenyl)-1,3-thiazole-4-carboxylic acid
SMILES C1=CC(=C(C(=C1)Cl)Cl)C2=NC(=CS2)C(=O)O

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 274.13
Percent Purity 97%
Chemical Name or Material 2-(2, 3-Dichlorophenyl)-1, 3-thiazole-4-carboxylic acid
Hazard Category GHS Signal Word: Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.