Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3-Trimethylbutane, MP Biomedicals

Catalog No. ICN219417 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN219417 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN219417 Supplier MP Biomedicals, Inc Supplier No. 0521941701

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 464-06-2
Molecular Formula C7H16
Molecular Weight (g/mol) 100.205
InChI Key ZISSAWUMDACLOM-UHFFFAOYSA-N
Synonym triptan, triptane, butane, 2,2,3-trimethyl, unii-40v943jdgr, 2,3-trimethylbutane, butane,2,3-trimethyl, 2,2,3-trimethyl-butane, acmc-1ahi7, butane,2,2,3-trimethyl, 2,3,3-trimethylbutan-2-yl
PubChem CID 10044
IUPAC Name 2,2,3-trimethylbutane
SMILES CC(C)C(C)(C)C

Specifications

Quantity 10 g
Formula Weight 100.21
Chemical Name or Material 2, 2, 3-Trimethylbutane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.