Learn More
Sigma Organic Chemistry 2,2,4,4,6,6,8,8,10,10,12,12-Dodecamethylcyclohexasiloxane | 250MG | 540-97-6 | MFCD00144215
Supplier: Sigma Organic Chemistry 796859250MG

2,2,4,4,6,6,8,8,10,10,12,12-Dodecamethylcyclohexasiloxane, 250MG
About This Item:
Linear Formula: C12H36O6Si6
Storage: room temp
EINECS Number: 208-762-8
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.