Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,4,4,6,8,8-Heptamethylnonane, 98.3%, MP Biomedicals™

Catalog No. ICN15732280 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15732280 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15732280 Supplier MP Biomedicals, Inc Supplier No. 0215732280
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4393-09-3
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.21
InChI Key LVMPWFJVYMXSNY-UHFFFAOYSA-N
Synonym 2,3-dimethoxybenzylamine, 2,3-dimethoxyphenyl methanamine, 1-2,3-dimethoxyphenyl methanamine, benzenemethanamine, 2,3-dimethoxy, 2,3-dimethoxy-benzylamine, 2,3-dimethoxy benzylamine, pubchem7399, 2,3-dimethoxybenzyl amine, acmc-2097fc
PubChem CID 78106
IUPAC Name (2,3-dimethoxyphenyl)methanamine
SMILES COC1=CC=CC(=C1OC)CN

Specifications

Color Colorless
Assay Percent Range 98.3%
Quantity 100 mL
Formula Weight 226.45
Physical Form Liquid
Chemical Name or Material 2,2,4,4,6,8,8-Heptamethylnonane
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.