Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,4,4-Tetramethylpentane 99.0+%, TCI America™
SDP

Catalog No. T170425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T170425ML 25 mL
T17045ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T170425ML Supplier TCI America Supplier No. T170425ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1070-87-7
Molecular Formula C9H20
Molecular Weight (g/mol) 128.26
MDL Number MFCD00060873
InChI Key GUMULFRCHLJNDY-UHFFFAOYSA-N
PubChem CID 14058
IUPAC Name 2,2,4,4-tetramethylpentane
SMILES CC(C)(C)CC(C)(C)C

Specifications

Color Colorless
Boiling Point 122°C
Quantity 25 mL
UN Number 1920
Synonym di-tert-butylmethane, pentane, 2,2,4,4-tetramethyl, unii-7js6eal3ee, 7js6eal3ee, pentane, 2,2,4,4-tetramethyl-8ci 9ci, 2,4,4-tetramethylpentane, acmc-2098nd, pentane,2,4,4-tetramethyl, 2,2,4,4,-tetramethylpentane, pentane,2,2,4,4-tetramethyl
Formula Weight 128.26
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,4,4-Tetramethylpentane
EINECSNumber (2)-0009
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.