Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,4,6,6-Pentamethylheptane 98.0+%, TCI America™
SDP

Catalog No. P004725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P004725ML 25 mL
P00475ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P004725ML Supplier TCI America Supplier No. P004725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13475-82-6
Molecular Formula C12H26
Molecular Weight (g/mol) 170.34
MDL Number MFCD00026334
InChI Key VKPSKYDESGTTFR-UHFFFAOYSA-N
PubChem CID 26058
ChEBI CHEBI:77509
IUPAC Name 2,2,4,6,6-pentamethylheptane
SMILES CC(CC(C)(C)C)CC(C)(C)C

Specifications

Color Colorless
Boiling Point 180°C
Quantity 25 mL
UN Number 2286
Synonym heptane, 2,2,4,6,6-pentamethyl, heptane,2,2,4,6,6-pentamethyl, permethyl 99a, acmc-1c04u, vkpskydesgttfr-uhfffaoysa, 2,2,4,6,6-pentamethyl-heptane
Formula Weight 170.34
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,4,6,6-Pentamethylheptane
EINECSNumber (2)-0010
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.