Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,4-Trimethylpentane, Analytical Standard, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018171 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8171 50 mL
11-101-9809 5 mL
11-101-9005 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-8171 Supplier MilliporeSigma Supelco Supplier No. 5903050ML
Add to Cart
Add to Cart

2,2,4-Trimethylpentane, a branched alkane, having a low tendency to auto-ignite, is used as a reference in determining the octane number of gasoline.

TRUSTED_SUSTAINABILITY

Specifications

CAS 540-84-1
Vapor Pressure 41 mmHg (21°C)
Vapor Density 3.9 (vs. air)
Density 0.692 g/mL (at 25°C)
Physical Form Neat
Linear Formula (CH3)2 CHCH2C(CH3)3
MDL Number MFCD00008943
Melting Point -107°C
Boiling Point 98°C to 99°C
Synonym Isooctane
Quantity 50 mL
Formula Weight 114.23
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.