Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid N-hydroxysuccinimide ester, 98+%

Catalog No. AAA1766701
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1766701 0.25 g
AAA1766703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1766701 Supplier Thermo Scientific Chemicals Supplier No. A1766701
Add to Cart

CAS: 37558-29-5 | C13H17N2O5

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37558-29-5
Molecular Formula C13H17N2O5
MDL Number MFCD00066369

Specifications

Melting Point 173°C to 177°C
Color Yellow
Quantity 0.25 g
Formula Weight 281.29
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material 2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid N-hydroxysuccinimide ester
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.