Learn More
Medchemexpress LLC 2-[2-(5-bromo-1H-indol-3-yl)ethyl]-3-[3-(1-methylethoxy)phenyl]-4-(3H)-quinazoline | 133040-77-4 | MFCD00907856 | 98.9% | 502.40 g/mol | C27H24BrN3O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10135225MG
Encompass_Preferred
LY 225910 is a selective antagonist of the cholecystokinin type 2 receptor (CCK2) used as a research tool to study CCK2-mediated signaling, opioid interactions such as morphine sensitization, and related pharmacology. Supplied as a solid research chemical, it is characterized by a brominated indole-quinazoline structure and analytical purity suitable for biochemical and in vivo assays.
  • Selective CCK2 receptor antagonist suitable for pharmacological studies.
  • High purity for research applications (≈98.9%).
  • Solid, white to off-white form for convenient handling and storage.
  • Stable under recommended storage conditions for prolonged shelf life.
  • Brominated indole-quinazoline scaffold supports receptor binding.
Catalog No. 50-002-12434